This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00498166_m1
See other IVD GE Assays ›
Gene Symbol:
Ivd
Gene Name:
isovaleryl coenzyme A dehydrogenase
Gene Aliases:
1300016K07Rik, 6720455E18Rik, AI463340
Chromosome Location:
Chr.2: 118861959 - 118882907 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00498166_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ivd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019826.3 NP_062800.1 1 - 2 168 80
GenBank mRNA AF225989.1 - 1 - 2 147 80
AK005024.1 - 1 - 2 187 80
AK077416.1 - 1 - 2 174 80
BC018325.1 - 1 - 2 158 4192286 80
BC027198.1 - 1 - 2 168 4237982 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001749.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ivd

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

leucine catabolic process
metabolic process
oxidation-reduction process
acyl-CoA dehydrogenase activity
isovaleryl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200013019.mm.1 NM_019826.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200013019.mm.1 NM_019826.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200013019.mm.1 TC0200001749.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200013017.mm.1 NM_019826.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200013017.mm.1 NM_019826.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200013017.mm.1 TC0200001749.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0