This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00496458_m1
See other ZFP346 GE Assays ›
Gene Symbol:
Zfp346
Gene Name:
zinc finger protein 346
Gene Aliases:
Jaz, Znf346
Chromosome Location:
Chr.13: 55105271 - 55135071 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00496458_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp346
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012017.2 NP_036147.1 3 - 4 407 62
GenBank mRNA AF083339.1 - 3 - 4 405 62
BC055075.1 - 3 - 4 401 30056643 62
CV562260.1 - 2 - 3 203 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000647.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp346

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of apoptotic process
nucleic acid binding
RNA binding
double-stranded RNA binding
protein binding
zinc ion binding
enzyme binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300004218.mm.1 NM_012017.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004218.mm.1 NM_012017.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004218.mm.1 TC1300000647.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0