This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00496413_m1
See other CREB3L1 GE Assays ›
Gene Symbol:
Creb3l1
Gene Name:
cAMP responsive element binding protein 3-like 1
Gene Aliases:
Oasis
Chromosome Location:
Chr.2: 91982328 - 92024170 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00496413_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Creb3l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011957.2 NP_036087.2 9 - 10 1202 87
GenBank mRNA AB017614.1 - 9 - 10 1164 87
AK142368.1 - 1203 87
BC016447.1 - 9 - 10 1177 4921584 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004133.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Creb3l1

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to unfolded protein
regulation of ossification
endoplasmic reticulum unfolded protein response
positive regulation of transcription from RNA polymerase II promoter
extracellular matrix constituent secretion
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
cAMP response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200034663.mm.1 NM_011957.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034663.mm.1 NM_011957.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034663.mm.1 TC0200004133.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200034664.mm.1 NM_011957.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034664.mm.1 NM_011957.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034664.mm.1 TC0200004133.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header