This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495444_m1
See other HSPA4L GE Assays ›
Gene Symbol:
Hspa4l
Gene Name:
heat shock protein 4 like
Gene Aliases:
94kDa, AI461691, APG-1, Osp94
Chromosome Location:
Chr.3: 40744355 - 40796103 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495444_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hspa4l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011020.3 NP_035150.3 4 - 5 491 96
GenBank mRNA AB001926.1 - 4 - 5 457 96
AK144225.1 - 4 - 5 528 96
AK163459.1 - 5 - 6 663 96
BC012712.1 - 4 - 5 560 4190161 96
BC057002.1 - 4 - 5 491 5702602 96
BC110662.1 - 4 - 5 511 4511154 96
D49482.1 - 4 - 5 485 96
U23921.1 - 4 - 5 608 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000307.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hspa4l

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to unfolded protein
nucleotide binding
protein binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300002215.mm.1 NM_011020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002215.mm.1 NM_011020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300002215.mm.1 TC0300000307.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300002217.mm.1 NM_011020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002217.mm.1 NM_011020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300002217.mm.1 TC0300000307.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300002212.mm.1 NM_011020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002212.mm.1 NM_011020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300002212.mm.1 TC0300000307.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300002213.mm.1 NM_011020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300002213.mm.1 NM_011020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300002213.mm.1 TC0300000307.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header