This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494840_g1
See other GTF2I GE Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6
Chromosome Location:
Chr.5: 134237832 - 134314760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494840_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080746.2 NP_001074215.1 32 - 33 3281 66
NM_001080747.2 NP_001074216.1 31 - 32 3218 66
NM_001080748.2 NP_001074217.1 30 - 31 3161 66
NM_001080749.2 NP_001074218.1 30 - 31 3173 66
NM_010365.4 NP_034495.2 31 - 32 3224 66
GenBank mRNA AF017085.1 - 32 - 33 2925 66
AF043219.1 - 30 - 31 2845 66
AF043220.1 - 29 - 30 2782 66
AK145698.1 - 22 - 23 2109 66
AK147201.1 - 30 - 31 3131 66
AK183312.1 - 3 - 4 101 66
AK211276.1 - 3 - 4 101 66
AY030290.1 - 31 - 32 2911 66
AY030291.1 - 31 - 32 2917 66
AY030292.1 - 30 - 31 2854 66
AY030293.1 - 32 - 33 2974 66
BC047387.1 - 30 - 31 2820 66
BC053044.1 - 32 - 33 3250 5704935 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003369.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2i

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
embryo development
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
DNA binding
protein binding
mitogen-activated protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030285.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030285.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030285.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030285.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030285.mm.1 NM_010365.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500030285.mm.1 TC0500003369.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0