This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00493960_m1
See other ZRSR2 GE Assays ›
Gene Symbol:
Zrsr2
Gene Name:
zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2
Gene Aliases:
35kDa, 5031411E02Rik, A230052C13Rik, C77286, U2af1-rs2, URP
Chromosome Location:
Chr.X: 163935443 - 163958820 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00493960_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zrsr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009453.3 NP_033479.2 8 - 9 1126 81
GenBank mRNA CN700359.1 - 1 - 2 347 81
D45205.1 - 8 - 9 1049 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003323.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zrsr2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal complex assembly
mRNA splicing, via spliceosome
mRNA processing
RNA splicing
nucleotide binding
nucleic acid binding
RNA binding
pre-mRNA 3'-splice site binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00020683.mm.1 NM_009453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020683.mm.1 NM_009453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020683.mm.1 TC0X00003323.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0