This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00491700_m1
See other SIGIRR GE Assays ›
Gene Symbol:
Sigirr
Gene Name:
single immunoglobulin and toll-interleukin 1 receptor (TIR) domain
Gene Aliases:
AI256711, TIR8
Chromosome Location:
Chr.7: 141091175 - 141100610 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00491700_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sigirr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023059.3 NP_075546.2 2 - 3 603 84
GenBank mRNA AF113795.1 - 1 - 2 120 84
AF239957.1 - 1 - 2 114 84
AK133518.1 - 2 - 3 228 84
AK145889.1 - 2 - 3 606 84
BC010806.1 - 1 - 2 125 4240425 84
BC094069.1 - 2 - 3 209 4974962 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004519.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF113795.1,AF239957.1,AK133518.1,AK145889.1,BC010806.1,BC094069.1,NM_023059.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sigirr

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

negative regulation of cytokine-mediated signaling pathway
acute-phase response
signal transduction
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of chemokine biosynthetic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700037921.mm.1 NM_023059.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037921.mm.1 NM_023059.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700037921.mm.1 TC0700004519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700037924.mm.1 NM_023059.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700037924.mm.1 NM_023059.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700037924.mm.1 TC0700004519.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0