This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00490177_m1
See other VCAN GE Assays ›
Gene Symbol:
Vcan
Gene Name:
versican
Gene Aliases:
5430420N07Rik, 9430051N09, Cspg2, DPEAAE, NG2, PG-M, PG-M(V0), PG-M(V1), hdf
Chromosome Location:
Chr.13: 89655309 - 89742694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00490177_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vcan
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081249.1 NP_001074718.1 11 - 12 9978 77
NM_001134474.1 NP_001127946.1 10 - 11 4761 77
NM_001134475.1 NP_001127947.1 9 - 10 1881 77
NM_019389.2 NP_062262.2 10 - 11 7098 77
GenBank mRNA AK045314.1 - 4 - 5 3433 77
D16263.1 - 9 - 10 6670 77
D28599.1 - 11 - 12 9722 77
D32040.1 - 9 - 10 1613 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vcan

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
osteoblast differentiation
cell adhesion
central nervous system development
heart development
glial cell migration
regulation of phosphoprotein phosphatase activity
calcium ion binding
hyaluronic acid binding
protein phosphatase binding
carbohydrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0