Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500011691.mm.1 |
NM_016667.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011691.mm.1 |
NM_016667.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011691.mm.1 |
TC1500001544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011692.mm.1 |
NM_016667.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011692.mm.1 |
NM_016667.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011692.mm.1 |
TC1500001544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011688.mm.1 |
TC1500001544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011690.mm.1 |
NM_016667.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011690.mm.1 |
NM_016667.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011690.mm.1 |
TC1500001544.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011693.mm.1 |
NM_016667.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011693.mm.1 |
NM_016667.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011693.mm.1 |
TC1500001544.mm.1 |
MTA TC |