This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489444_mH
See other SLC25A13 GE Assays ›
Gene Symbol:
Slc25a13
Gene Name:
solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13
Gene Aliases:
AI785475, Ctrn
Chromosome Location:
Chr.6: 6041218 - 6217233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489444_mH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177572.1 NP_001171043.1 15 - 16 1756 96
NM_015829.3 NP_056644.1 15 - 16 1759 96
GenBank mRNA AF164632.1 - 15 - 16 1704 96
AK002829.1 - 15 - 16 1634 96
AK012670.1 - 15 - 16 1647 96
AK029020.1 - 15 - 16 1687 96
AK150496.1 - 15 - 16 1701 96
AK151180.1 - 15 - 16 1701 96
AK152237.1 - 15 - 16 1701 96
BC016571.1 - 9 - 10 893 3600909 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001813.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a13

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
transport
aspartate transport
L-glutamate transport
malate-aspartate shuttle
cellular respiration
response to calcium ion
transmembrane transport
L-glutamate transmembrane transport
L-aspartate transmembrane transport
L-glutamate transmembrane transporter activity
calcium ion binding
L-aspartate transmembrane transporter activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600014382.mm.1 NM_001177572.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014382.mm.1 NM_001177572.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014382.mm.1 NM_015829.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600014382.mm.1 NM_015829.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600014382.mm.1 TC0600001813.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0