This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486720_m1
See other CAR6 GE Assays ›
Gene Symbol:
Car6
Gene Name:
carbonic anhydrase 6
Gene Aliases:
Ca6, DOC1
Chromosome Location:
Chr.4: 150187015 - 150201135 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486720_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Car6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009802.2 NP_033932.2 4 - 5 516 94
GenBank mRNA AF079834.1 - 3 - 4 606 94
AF079835.1 - 4 - 5 543 94
AK132599.1 - 4 - 5 517 94
AK161216.1 - 4 - 5 512 94
BC006726.1 - 4 - 5 603 94
BC018582.1 - 4 - 5 493 94
BC036270.1 - 4 - 5 499 94
BC039657.1 - 4 - 5 501 94
BC046495.1 - 3 - 4 292 94
BC049973.1 - 4 - 5 514 94
BC055437.1 - 4 - 5 502 4190566 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004062.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Car6

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
one-carbon metabolic process
molecular_function
carbonate dehydratase activity
zinc ion binding
lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400033768.mm.1 NM_009802.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400033768.mm.1 NM_009802.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400033768.mm.1 TC0400004062.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0