This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485814_m1
See other POLR1B GE Assays ›
Gene Symbol:
Polr1b
Gene Name:
polymerase (RNA) I polypeptide B
Gene Aliases:
128kDa, D630020H17Rik, RPA116, RPA135, RPA2, Rpo1-2
Chromosome Location:
Chr.2: 129100970 - 129128060 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485814_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009086.2 NP_033112.2 5 - 6 851 123
GenBank mRNA AK052672.1 - 5 - 6 821 123
AK083732.1 - 5 - 6 833 123
AK143501.1 - 5 - 6 788 123
AK144616.1 - 5 - 6 838 123
AK156712.1 - 5 - 6 842 123
BC060656.1 - 5 - 6 781 30355130 123
U58280.1 - 5 - 6 815 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001947.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr1b

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
embryo implantation
rRNA transcription
embryo development
nucleologenesis
RNA polymerase I activity
DNA binding
DNA-directed RNA polymerase activity
protein binding
transferase activity
nucleotidyltransferase activity
ribonucleoside binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200015340.mm.1 NM_009086.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200015340.mm.1 NM_009086.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200015340.mm.1 TC0200001947.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0