Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700007926.mm.1 |
NM_008650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007926.mm.1 |
NM_008650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007926.mm.1 |
TC1700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007924.mm.1 |
NM_008650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007924.mm.1 |
NM_008650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007924.mm.1 |
TC1700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007925.mm.1 |
NM_008650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007925.mm.1 |
NM_008650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007925.mm.1 |
TC1700000819.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007923.mm.1 |
NM_008650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007923.mm.1 |
NM_008650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007923.mm.1 |
TC1700000819.mm.1 |
MTA TC |