This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00482814_s1
See other PPP1R2-PS9 GE Assays ›
Gene Symbol:
Ppp1r2-ps9
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 2, pseudogene 9
Gene Aliases:
4930403L05Rik
Chromosome Location:
Chr.X: 15110586 - 15111489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00482814_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r2-ps9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033171.1 1 745 110
GenBank mRNA AK015065.1 - 1 750 110
AK015311.1 - 1 722 110
BC100448.1 - 1 722 30918995 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001883.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase inhibitor phosphatase modulator

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 BC132484.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 BC132486.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 NR_033171.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 AK015065.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 AK015311.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 BC132484.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 BC132486.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 NR_033171.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 AK015065.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210403 AK015311.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 BC132484.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 BC132486.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 NR_033171.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 AK015065.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 AK015311.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 BC132484.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 BC132486.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 NR_033171.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 AK015065.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P210403 AK015311.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 BC132484.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 BC132486.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 NR_033171.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 AK015065.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 AK015311.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 BC132484.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 BC132486.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 NR_033171.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 AK015065.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P210403 AK015311.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10603741 - -
Affymetrix Mouse Gene 1.0 ST Array 10603740 - -
Affymetrix Mouse Gene 1.0 ST Array 10603739 - -
Affymetrix HT MG-430 PM Array Plate 1419541_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10603738 - -
Mouse Genome 430 2.0 Array 1419541_at - -
Mouse Genome 430 2.0 Array 1419541_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67395_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 67395_at - -
Affymetrix Mouse Clariom S Array PSR0X00012048.mm.1 NR_033171.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00012048.mm.1 TC0X00001883.mm.1 MTA TC
Mouse Expression Set 430 1419541_PM_at - -
Mouse Expression Set 430 1419541_at - -
Murine Genome U74v2 Set 169155_f_at - -
Murine Genome U74v2 Set 169155_PM_f_at - -
Mouse Genome 430A 2.0 Array 1419541_at - -
Mouse Genome 430A 2.0 Array 1419541_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419541_at - -
Affymetrix HT Mouse Genome 430A Array 1419541_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 586970 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673241 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2673241 - -
MouseRef-8 v2 Expression BeadChip ILMN_2803156 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803156 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0