This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480342_m1
See other NSMF GE Assays ›
Gene Symbol:
Nsmf
Gene Name:
NMDA receptor synaptonuclear signaling and neuronal migration factor
Gene Aliases:
Jacob, Nelf
Chromosome Location:
Chr.2: 25054370 - 25062881 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480342_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nsmf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039386.1 NP_001034475.1 3 - 4 851 100
NM_001039387.1 NP_001034476.1 3 - 4 851 100
NM_001177654.1 NP_001171125.1 3 - 4 851 100
NM_001177655.1 NP_001171126.1 3 - 4 851 100
NM_020276.3 NP_064672.2 3 - 4 851 100
GenBank mRNA AF266508.2 - 3 - 4 817 100
AJ278902.1 - 3 - 4 629 100
AK004986.2 - 3 - 4 848 100
AK019560.1 - 3 - 4 818 100
AK053402.1 - 3 - 4 798 100
AK158803.1 - 4 - 5 650 100
AK165300.1 - 3 - 4 796 100
BC006642.1 - 3 - 4 809 3483536 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000395.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nsmf

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein dephosphorylation
regulation of neuron apoptotic process
regulation of neuronal synaptic plasticity
regulation of dendrite morphogenesis
cellular response to amino acid stimulus
cellular response to electrical stimulus
cellular response to gonadotropin stimulus
positive regulation of neuron migration
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001039386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001039386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001039387.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001039387.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001177654.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001177654.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001177655.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_001177655.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_020276.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 NM_020276.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200002133.mm.1 TC0200000395.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0