This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480118_m1
See other ARPC3 GE Assays ›
Gene Symbol:
Arpc3
Gene Name:
actin related protein 2/3 complex, subunit 3
Gene Aliases:
1110006A04Rik, 21kDa, AA408672, AI788639, p21-Ar, p21Arc
Chromosome Location:
Chr.5: 122391928 - 122406178 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480118_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arpc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019824.3 NP_062798.1 4 - 5 320 91
GenBank mRNA AB038243.1 - 3 - 4 256 91
AK003485.1 - 4 - 5 351 91
AK005437.1 - 4 - 5 339 91
AK171651.1 - 4 - 5 335 91
BC013618.1 - 4 - 5 320 4013537 91
BC054440.1 - 4 - 5 287 6395280 91
BC055036.1 - 4 - 5 287 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001392.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arpc3

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of actin filament polymerization
Arp2/3 complex-mediated actin nucleation
actin binding
structural constituent of cytoskeleton
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500012321.mm.1 NM_019824.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500012321.mm.1 TC0500001392.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0