This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479914_m1
See other HSD17B12 GE Assays ›
Gene Symbol:
Hsd17b12
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 12
Gene Aliases:
2610510O05Rik, AI172963, KIK-I, Kik1
Chromosome Location:
Chr.2: 94032697 - 94157909 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479914_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd17b12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019657.4 NP_062631.1 4 - 5 436 90
GenBank mRNA AF064635.1 - 4 - 5 415 90
AK078841.1 - 4 - 5 437 90
AK081869.1 - 4 - 5 430 90
AK082715.1 - 4 - 5 430 90
AK088521.1 - 4 - 5 430 90
AK150849.1 - 4 - 5 431 90
AK152489.1 - 4 - 5 431 90
AK166027.1 - 4 - 5 430 90
AK167908.1 - 4 - 5 430 90
BC037620.1 - 3 - 4 353 5375831 90
BC090659.1 - 4 - 5 406 30675153 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004169.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC037620.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd17b12

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid biosynthetic process
metabolic process
positive regulation of cell-substrate adhesion
extracellular matrix organization
oxidation-reduction process
fibronectin binding
estradiol 17-beta-dehydrogenase activity
collagen binding
heparin binding
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200035099.mm.1 NM_019657.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035099.mm.1 NM_019657.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200035099.mm.1 TC0200004169.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200035100.mm.1 NM_019657.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200035100.mm.1 NM_019657.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200035100.mm.1 TC0200004169.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0