This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479582_g1
See other PDE3A GE Assays ›
Gene Symbol:
Pde3a
Gene Name:
phosphodiesterase 3A, cGMP inhibited
Gene Aliases:
A930022O17Rik, C87899
Chromosome Location:
Chr.6: 141249269 - 141506863 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479582_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018779.1 NP_061249.1 3 - 4 1580 67
GenBank mRNA AF099187.1 - 3 - 4 1580 67
AK004638.1 - 3 - 4 885 67
AK052374.1 - 3 - 4 858 67
AK164864.1 - 3 - 4 360 67
BC138265.1 - 3 - 4 1538 8861284 67
BC138266.1 - 3 - 4 1538 8861285 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001664.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde3a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
cAMP catabolic process
signal transduction
cAMP-mediated signaling
cGMP-mediated signaling
regulation of meiotic nuclear division
response to drug
negative regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of vascular permeability
positive regulation of oocyte development
cellular response to cGMP
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
cAMP binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600013274.mm.1 NM_018779.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013274.mm.1 NM_018779.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013274.mm.1 TC0600001664.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0