This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00473008_m1
See other SIL1 GE Assays ›
Gene Symbol:
Sil1
Gene Name:
endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae)
Gene Aliases:
1810057E01Rik, AI042831, wz
Chromosome Location:
Chr.18: 35266396 - 35499052 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00473008_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sil1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030749.2 NP_109674.2 2 - 3 243 73
GenBank mRNA AJ297884.1 - 2 - 3 184 73
AK170415.1 - 2 - 3 196 73
BC016119.1 - 2 - 3 220 4489217 73
BC016466.1 - 2 - 3 206 3582798 73
CX219879.1 - 2 - 3 243 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001171.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sil1

Gene Ontology Categories:

Function(s)

transport
protein transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800008619.mm.1 NM_030749.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008619.mm.1 NM_030749.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800008619.mm.1 TC1800001171.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0