Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019263.mm.1 |
NM_001006664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019263.mm.1 |
TC0200002395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019268.mm.1 |
NM_001006664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019268.mm.1 |
NM_013510.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019268.mm.1 |
TC0200002395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019262.mm.1 |
NM_001006664.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019262.mm.1 |
TC0200002395.mm.1 |
MTA TC |