This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00463513_m1
See other ALDH1L2 GE Assays ›
Gene Symbol:
Aldh1l2
Gene Name:
aldehyde dehydrogenase 1 family, member L2
Gene Aliases:
D330038I09Rik, mtFDH
Chromosome Location:
Chr.10: 83487283 - 83534146 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00463513_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh1l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153543.2 NP_705771.2 15 - 16 1992 66
GenBank mRNA AK052361.1 - 15 - 16 1883 66
AK086413.1 - 15 - 16 1936 66
AK141954.1 - 15 - 16 1953 66
BC034531.1 - 15 - 16 1914 4188055 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002583.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh1l2

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

one-carbon metabolic process
metabolic process
biosynthetic process
10-formyltetrahydrofolate catabolic process
oxidation-reduction process
catalytic activity
aldehyde dehydrogenase (NAD) activity
methyltransferase activity
formyltetrahydrofolate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
hydroxymethyl-, formyl- and related transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000019461.mm.1 NM_153543.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000019461.mm.1 NM_153543.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000019461.mm.1 TC1000002583.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000019460.mm.1 NM_153543.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000019460.mm.1 NM_153543.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000019460.mm.1 TC1000002583.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0