This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00462670_g1
See other AGO4 GE Assays ›
Gene Symbol:
Ago4
Gene Name:
argonaute RISC catalytic subunit 4
Gene Aliases:
5730550L01Rik, AI481660, Eif2c4
Chromosome Location:
Chr.4: 126489487 - 126533588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00462670_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ago4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153177.3 NP_694817.2 14 - 15 2292 100
GenBank mRNA AB081474.1 - 14 - 15 2037 100
AK030018.1 - 14 - 15 2308 100
AK032475.1 - 3 - 4 617 100
AK129395.2 - 10 - 11 1474 100
BC096023.1 - 14 - 15 2074 30607807 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003545.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ago4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA catabolic process
regulation of translation
RNA secondary structure unwinding
gene silencing by RNA
pre-miRNA processing
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
miRNA loading onto RISC involved in gene silencing by miRNA
nucleic acid binding
RNA binding
double-stranded RNA binding
single-stranded RNA binding
miRNA binding
pre-miRNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400028875.mm.1 NM_153177.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028875.mm.1 NM_153177.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400028875.mm.1 TC0400003545.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0