This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461365_g1
See other PRR5 GE Assays ›
Gene Symbol:
Prr5
Gene Name:
proline rich 5 (renal)
Gene Aliases:
AU043908, Arhgap8, C030017C09Rik, C78947, Protor-1, Protor1
Chromosome Location:
Chr.15: 84663171 - 84706409 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461365_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prr5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146061.4 NP_666173.4 3 - 4 452 84
GenBank mRNA AF482997.1 - 3 - 4 447 84
AK046057.1 - 4 - 5 269 84
AK081231.1 - 3 - 4 448 84
BC024991.1 - 4 - 5 336 5065129 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000791.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prr5

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
cell cycle
positive regulation of phosphatidylinositol 3-kinase signaling
actin cytoskeleton organization
positive regulation of cell migration
positive regulation of GTPase activity
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500006231.mm.1 NM_146061.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006231.mm.1 NM_146061.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500006231.mm.1 TC1500000791.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500006232.mm.1 NM_146061.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006232.mm.1 NM_146061.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500006232.mm.1 TC1500000791.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0