This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00458902_g1
See other LUC7L3 GE Assays ›
Gene Symbol:
Luc7l3
Gene Name:
LUC7-like 3 (S. cerevisiae)
Gene Aliases:
3300001P08Rik, Luc7a
Chromosome Location:
Chr.11: 94291139 - 94321983 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00458902_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Luc7l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026313.1 NP_080589.1 7 - 8 811 80
GenBank mRNA AK014362.1 - 7 - 8 804 80
AK078090.1 - 7 - 8 858 80
AK084714.1 - 2 - 3 198 80
AK151228.1 - 7 - 8 859 80
AK151839.1 - 7 - 8 859 80
BC009092.1 - 7 - 8 811 3157532 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003559.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Luc7l3

Gene Ontology Categories:

Function(s) Process(es)

mRNA splice site selection
mRNA processing
RNA splicing
DNA binding
mRNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100032440.mm.1 NM_026313.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032440.mm.1 NM_026313.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032440.mm.1 TC1100003559.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032438.mm.1 NM_026313.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032438.mm.1 NM_026313.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032438.mm.1 TC1100003559.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0