This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00456440_m1
See other ADAM17 GE Assays ›
Gene Symbol:
Adam17
Gene Name:
a disintegrin and metallopeptidase domain 17
Gene Aliases:
CD156b, Tace
Chromosome Location:
Chr.12: 21323509 - 21373632 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00456440_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adam17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277266.1 NP_001264195.1 17 - 18 2253 70
NM_001291871.1 NP_001278800.1 16 - 17 2215 70
NM_009615.6 NP_033745.4 16 - 17 2196 70
NR_102380.1 13 - 14 1839 70
NR_120376.1 16 - 17 2219 70
GenBank mRNA AB021709.1 - 16 - 17 2157 70
AJ007365.1 - 16 - 17 1996 70
AK137183.1 - 8 - 9 3305 70
AK137693.1 - 16 - 17 2103 70
AK139471.1 - 16 - 17 2102 70
AK142441.1 - 16 - 17 2155 70
AK144161.1 - 17 - 18 2183 70
AK149288.1 - 16 - 17 2099 70
BC026052.1 - 13 - 14 1735 70
BC048792.1 - 16 - 17 2103 70
BC094655.1 - 16 - 17 2110 5705503 70
BC114999.1 - 16 - 17 2056 6506501 70
BC138416.1 - 16 - 17 2075 8861436 70
BC138420.1 - 16 - 17 2075 8861440 70
BC145270.1 - 16 - 17 2075 9053824 70
U69613.1 - 16 - 17 2064 70
U69614.1 - 16 - 17 2171 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001490.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adam17

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
positive regulation of protein phosphorylation
germinal center formation
proteolysis
membrane protein ectodomain proteolysis
cell adhesion
epidermal growth factor receptor signaling pathway
Notch signaling pathway
Notch receptor processing
positive regulation of cell proliferation
positive regulation of T cell chemotaxis
B cell differentiation
positive regulation of cell growth
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
response to lipopolysaccharide
positive regulation of chemokine production
regulation of mast cell apoptotic process
T cell differentiation in thymus
cell adhesion mediated by integrin
epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
response to drug
negative regulation of apoptotic process
positive regulation of epidermal growth factor-activated receptor activity
spleen development
cell motility
defense response to Gram-positive bacterium
PMA-inducible membrane protein ectodomain proteolysis
positive regulation of cellular component movement
response to high density lipoprotein particle
metalloendopeptidase activity
Notch binding
interleukin-6 receptor binding
integrin binding
protein binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
SH3 domain binding
PDZ domain binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200010445.mm.1 NM_001277266.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200010445.mm.1 NM_009615.6 Refseq
Affymetrix Mouse Clariom S Array PSR1200010445.mm.1 NR_102380.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200010445.mm.1 TC1200001490.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header