This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00455832_mH
See other RPS6KA3 GE Assays ›
Gene Symbol:
Rps6ka3
Gene Name:
ribosomal protein S6 kinase polypeptide 3
Gene Aliases:
C130006E23, MAPKAPK-1b, MPK-9, Rsk2, S6K-alpha3, p90RSK3, pp90RSK2
Chromosome Location:
Chr.X: 159255782 - 159368244 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00455832_mH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps6ka3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_148945.2 NP_683747.1 18 - 19 2097 110
GenBank mRNA AK154778.1 - 18 - 19 2009 110
AY083469.1 - 18 - 19 1759 110
BC038683.1 - 2 - 3 183 110
BC150156.1 - 18 - 19 1762 110
BC150478.1 - 18 - 19 1762 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001612.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps6ka3

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

toll-like receptor signaling pathway
regulation of translation
protein phosphorylation
cell cycle
phosphorylation
response to lipopolysaccharide
intracellular signal transduction
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription from RNA polymerase II promoter
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ribosomal protein S6 kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00010138.mm.1 NM_148945.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00010138.mm.1 TC0X00001612.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0