This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453910_m1
See other TMEM132A GE Assays ›
Gene Symbol:
Tmem132a
Gene Name:
transmembrane protein 132A
Gene Aliases:
6720481D13Rik, Hspa5bp1, Orai1, R74613
Chromosome Location:
Chr.19: 10857829 - 10869935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453910_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem132a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133804.2 NP_598565.2 4 - 5 1003 75
GenBank mRNA AK032956.1 - 4 - 5 1004 75
AK036651.1 - 4 - 5 1003 75
AK048556.1 - 4 - 5 1004 75
AK164646.1 - 4 - 5 1005 75
AK173207.1 - 4 - 5 991 75
BC006896.1 - 4 - 5 955 3599652 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001115.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem132a

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of programmed cell death
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900010467.mm.1 NM_133804.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900010467.mm.1 NM_133804.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900010467.mm.1 TC1900001115.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0