This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453676_m1
See other MVP GE Assays ›
Gene Symbol:
Mvp
Gene Name:
major vault protein
Gene Aliases:
2310009M24Rik, LRP, VAULT1
Chromosome Location:
Chr.7: 126986860 - 127014594 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453676_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mvp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080638.3 NP_542369.2 1 - 2 83 70
GenBank mRNA AF210456.1 - 1 - 2 85 70
AK088046.1 - 1 - 2 68 70
AK152534.1 - 1 - 2 58 70
AK167899.1 - 1 - 2 59 70
AK168100.1 - 1 - 2 59 70
BB851648.1 - 1 - 2 84 70
BC006709.1 - 1 - 2 48 3584439 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004660.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF210456.1,AK088046.1,AK152534.1,AK167899.1,AK168100.1,BC006709.1,NM_080638.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mvp

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
negative regulation of signaling
negative regulation of protein autophosphorylation
ERBB signaling pathway
negative regulation of protein tyrosine kinase activity
protein activation cascade
protein kinase binding
protein phosphatase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700039195.mm.1 NM_080638.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700039195.mm.1 NM_080638.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700039195.mm.1 TC0700004660.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700036121.mm.1 NM_080638.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036121.mm.1 NM_080638.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036121.mm.1 TC0700004660.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700036124.mm.1 NM_080638.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036124.mm.1 NM_080638.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036124.mm.1 TC0700004660.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700036123.mm.1 NM_080638.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036123.mm.1 NM_080638.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700036123.mm.1 TC0700004660.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 722366 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0