This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453244_m1
See other OGFR GE Assays ›
Gene Symbol:
Ogfr
Gene Name:
opioid growth factor receptor
Gene Aliases:
2010013E17Rik
Chromosome Location:
Chr.2: 180589407 - 180595837 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453244_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogfr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031373.3 NP_113550.3 3 - 4 359 102
GenBank mRNA AF303894.1 - 3 - 4 307 102
AK008230.1 - 3 - 4 324 102
AK149146.1 - 3 - 4 306 102
AK150115.1 - 3 - 4 292 102
BC003875.1 - 3 - 4 290 3495186 102
BC049965.1 - 3 - 4 305 3155091 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002796.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogfr

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

regulation of growth
receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022879.mm.1 NM_031373.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022879.mm.1 NM_031373.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200022879.mm.1 TC0200002796.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0