This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453207_s1
See other UCN3 GE Assays ›
Gene Symbol:
Ucn3
Gene Name:
urocortin 3
Gene Aliases:
-
Chromosome Location:
Chr.13: 3940688 - 3945349 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453207_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucn3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031250.5 NP_112540.2 2 778 59
GenBank mRNA AK148451.1 - 2 779 59
BC119433.1 - 1 263 40041175 59
BC119434.1 - 1 260 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001375.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucn3

Gene Ontology Categories:

Function(s) Process(es)

response to stress
adenylate cyclase-activating G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
digestion
cellular response to nutrient levels
positive regulation of insulin secretion
positive regulation of membrane potential
G-protein coupled receptor binding
hormone activity
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone receptor 2 binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046403 AK148451.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046403 NM_031250.5 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960087 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461614 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174096 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105954 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046403 AK148451.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046403 NM_031250.5 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P342327 AK148451.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P342327 BC119433.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P342327 BC119434.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P342327 NM_031250.5 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105954 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02619568 - -
Whole Mouse Genome G4122A A_51_P342327 AK148451.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P342327 BC119433.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P342327 BC119434.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P342327 NM_031250.5 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107170 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105954 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10878788 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10959606 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10959610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790726 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790685 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790740 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790722 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790753 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790734 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790625 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790713 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790680 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790758 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790726 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10683145 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790753 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790722 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790760 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790694 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790734 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10967971 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790741 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790732 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790712 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790716 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790725 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790740 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790704 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790746 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10604074 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790677 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11067643 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790685 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10683142 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790683 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10476551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790750 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790688 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790695 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790709 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790713 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790745 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790718 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790729 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790675 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10898907 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790682 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10556517 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790739 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10881300 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10959612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790702 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10605001 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10898905 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790764 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790691 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790698 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790761 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790768 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790762 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790756 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790744 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790763 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790700 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790690 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10630984 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10847753 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10803237 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790738 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28790680 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10407432 - -
Affymetrix Mouse Gene 1.0 ST Array 10407433 - -
Affymetrix Mouse Gene 1.0 ST Array 10407434 - -
Affymetrix HT MG-430 PM Array Plate 1441439_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10407431 - -
Mouse Genome 430 2.0 Array 1450607_s_at - -
Mouse Genome 430 2.0 Array 1450607_PM_s_at - -
Mouse Genome 430 2.0 Array 1441439_at - -
Mouse Genome 430 2.0 Array 1441439_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 83428_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 83428_at - -
Affymetrix Mouse Clariom S Array PSR1300008622.mm.1 NM_031250.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300008622.mm.1 NM_031250.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300008622.mm.1 TC1300001375.mm.1 MTA TC
Mouse Expression Set 430 1450607_PM_s_at - -
Mouse Expression Set 430 1450607_s_at - -
Mouse Expression Set 430 1441439_PM_at - -
Mouse Expression Set 430 1441439_at - -
Mouse Genome 430A 2.0 Array 1450607_s_at - -
Mouse Genome 430A 2.0 Array 1450607_PM_s_at - -
Affymetrix HT Mouse Genome 430B Array 1441439_at - -
Affymetrix HT Mouse Genome 430B Array 1441439_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 538528 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4920020 - -
Illumina mouse-6 v1.0 expression beadchip 4920020 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004920020 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238143 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1238143 - -
mouse-6 v1.1 (Illumina) scl44315.2.7_1-S - -
Illumina Sentrix mouse-6.v1 bead array 4920020 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl44315.2.7_1-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920020 - -
MouseRef-8 v2 Expression BeadChip ILMN_1238143 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238143 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0