This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451513_m1
See other SLCO1B2 GE Assays ›
Gene Symbol:
Slco1b2
Gene Name:
solute carrier organic anion transporter family, member 1b2
Gene Aliases:
7330442B20Rik, OATP-C, OATP2, Oatp1b2, Oatp4, Slc21a10, Slc21a6, lst-1, mlst-1
Chromosome Location:
Chr.6: 141629452 - 141686646 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451513_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco1b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020495.1 NP_065241.1 14 - 15 1934 110
GenBank mRNA AB031959.1 - 14 - 15 1934 110
AF250912.1 - 6 - 7 1062 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001672.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco1b2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
drug transmembrane transport
oligopeptide transport
organic anion transport
bile acid and bile salt transport
drug transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
organic anion transmembrane transporter activity
bile acid transmembrane transporter activity
oligopeptide transporter activity
drug transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600013339.mm.1 NM_020495.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013339.mm.1 NM_020495.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013339.mm.1 TC0600001672.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0