This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451427_m1
See other SLC6A14 GE Assays ›
Gene Symbol:
Slc6a14
Gene Name:
solute carrier family 6 (neurotransmitter transporter), member 14
Gene Aliases:
1110007A17Rik, 9030613J17Rik, ATB0plus, CATB0plus
Chromosome Location:
Chr.X: 21714900 - 21742358 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451427_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc6a14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020049.4 NP_064433.3 6 - 7 1029 74
GenBank mRNA AB033285.1 - 6 - 7 787 74
AF161714.1 - 6 - 7 1029 74
AF320226.1 - 6 - 7 973 74
AK018553.1 - 6 - 7 888 74
BC129908.1 - 6 - 7 820 40111716 74
BC129909.1 - 6 - 7 820 40111717 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000229.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc6a14

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
amino acid transport
response to toxic substance
amine transport
transmembrane transport
amine transmembrane transporter activity
neurotransmitter:sodium symporter activity
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00001613.mm.1 NM_020049.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001613.mm.1 NM_020049.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001613.mm.1 TC0X00000229.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0