This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451224_m1
See other TRPC4AP GE Assays ›
Gene Symbol:
Trpc4ap
Gene Name:
transient receptor potential cation channel, subfamily C, member 4 associated protein
Gene Aliases:
4833429F06Rik, D2Ertd113e, Trrp4ap, mFLJ00177, truss
Chromosome Location:
Chr.2: 155634277 - 155692384 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451224_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trpc4ap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163452.1 NP_001156924.1 13 - 14 1670 85
NM_019828.2 NP_062802.2 13 - 14 1646 85
GenBank mRNA AF093589.1 - 9 - 10 1142 85
AF130458.1 - 13 - 14 1594 85
AK154263.1 - 13 - 14 1619 85
AK156047.1 - 13 - 14 1619 85
AK165259.1 - 13 - 14 1619 85
AK170085.1 - 13 - 14 1608 85
AK171404.1 - 13 - 14 1619 85
AK220239.1 - 13 - 14 1646 85
BC003931.1 - 6 - 7 606 85
BC033274.1 - 13 - 14 1598 3673725 85
BC057330.1 - 13 - 14 1628 6816155 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trpc4ap

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
phosphatase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0