This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451218_m1
See other TRPC4AP GE Assays ›
Gene Symbol:
Trpc4ap
Gene Name:
transient receptor potential cation channel, subfamily C, member 4 associated protein
Gene Aliases:
4833429F06Rik, D2Ertd113e, Trrp4ap, mFLJ00177, truss
Chromosome Location:
Chr.2: 155634277 - 155692384 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451218_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trpc4ap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163452.1 NP_001156924.1 7 - 8 943 58
NM_019828.2 NP_062802.2 7 - 8 943 58
GenBank mRNA AF093589.1 - 3 - 4 415 58
AF130458.1 - 7 - 8 891 58
AF509881.1 - 7 - 8 865 58
AK037501.1 - 2 - 3 374 58
AK076496.1 - 7 - 8 942 58
AK140799.1 - 6 - 7 631 58
AK154263.1 - 7 - 8 892 58
AK156047.1 - 7 - 8 892 58
AK165259.1 - 7 - 8 892 58
AK170085.1 - 7 - 8 881 58
AK171404.1 - 7 - 8 892 58
AK181057.1 - 1 - 2 199 58
AK220239.1 - 7 - 8 919 58
BC033274.1 - 7 - 8 871 3673725 58
BC057330.1 - 7 - 8 901 6816155 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004967.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trpc4ap

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042199.mm.1 NM_001163452.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042199.mm.1 NM_001163452.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042199.mm.1 NM_019828.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042199.mm.1 NM_019828.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042199.mm.1 TC0200004967.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0