This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00450164_m1
See other POLG2 GE Assays ›
Gene Symbol:
Polg2
Gene Name:
polymerase (DNA directed), gamma 2, accessory subunit
Gene Aliases:
-
Chromosome Location:
Chr.11: 106754821 - 106780010 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00450164_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015810.2 NP_056625.2 4 - 5 985 103
NR_027785.1 4 - 5 838 103
GenBank mRNA AF177202.1 - 4 - 5 890 103
AK041591.1 - 4 - 5 957 103
AK042777.1 - 4 - 5 796 103
AK164730.1 - 4 - 5 845 103
AK169977.1 - 4 - 5 1227 103
BC119131.1 - 4 - 5 914 8733880 103
BC144810.1 - 4 - 5 789 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003934.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polg2

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
DNA replication
mitochondrial DNA replication
DNA repair
respiratory electron transport chain
mitochondrial DNA metabolic process
mitochondrion morphogenesis
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100036890.mm.1 NM_015810.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036890.mm.1 NM_015810.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036890.mm.1 NR_027785.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036890.mm.1 TC1100003934.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100036892.mm.1 NM_015810.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100036892.mm.1 NM_015810.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036892.mm.1 NR_027785.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100036892.mm.1 TC1100003934.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0