This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00449764_g1
See other SUPT5 GE Assays ›
Gene Symbol:
Supt5
Gene Name:
suppressor of Ty 5
Gene Aliases:
AL033283, AL033321, AU019549, Spt5, Supt5h
Chromosome Location:
Chr.7: 28314891 - 28338804 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00449764_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Supt5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013676.1 NP_038704.1 25 - 26 2504 64
GenBank mRNA AK145117.1 - 25 - 26 2544 64
AK145465.1 - 17 - 18 1369 64
AK146055.1 - 25 - 26 2532 64
AK146453.1 - 25 - 26 2532 64
AK146583.1 - 24 - 25 2751 64
AK146650.1 - 25 - 26 2545 64
AK159786.1 - 26 - 27 2629 64
BC007132.1 - 25 - 26 2504 3496691 64
BC057449.1 - 25 - 26 2519 64
BC058598.1 - 24 - 25 2479 6306109 64
BC059849.1 - 17 - 18 2087 5696323 64
U88539.1 - 25 - 26 2531 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002694.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Supt5

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA-templated transcription, elongation
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
positive regulation of macroautophagy
regulation of DNA-templated transcription, elongation
negative regulation of DNA-templated transcription, elongation
positive regulation of DNA-templated transcription, elongation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of mRNA polyadenylation
chromatin binding
enzyme binding
poly(A) RNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700023535.mm.1 NM_013676.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023535.mm.1 NM_013676.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023535.mm.1 TC0700002694.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0