This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448524_m1
See other PXN GE Assays ›
Gene Symbol:
Pxn
Gene Name:
paxillin
Gene Aliases:
AW108311, AW123232, Pax
Chromosome Location:
Chr.5: 115506674 - 115555987 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448524_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pxn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011223.3 NP_035353.1 1 - 2 194 93
NM_133915.3 NP_598676.2 1 - 2 194 93
GenBank mRNA AF293882.1 - 1 - 2 119 93
AK148060.1 - 1 - 2 180 93
AK149933.1 - 1 - 2 159 93
AK167299.1 - 1 - 2 133 93
AK171226.1 - 1 - 2 183 93
AK171436.1 - 1 - 2 159 93
BC025493.1 - 1 - 2 152 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001264.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF293882.1,AK148060.1,AK149933.1,AK167299.1,AK171436.1,NM_011223.3,NM_133915.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pxn

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
movement of cell or subcellular component
cytoskeleton organization
cell adhesion
cell-matrix adhesion
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
regulation of cell shape
peptidyl-tyrosine phosphorylation
lamellipodium assembly
substrate adhesion-dependent cell spreading
positive regulation of protein kinase activity
focal adhesion assembly
branching morphogenesis of an epithelial tube
growth hormone receptor signaling pathway
integrin binding
protein binding
beta-catenin binding
zinc ion binding
vinculin binding
protein kinase binding
receptor signaling complex scaffold activity
protein complex binding
metal ion binding
BH4 domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500010903.mm.1 NM_011223.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010903.mm.1 NM_133915.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010903.mm.1 TC0500001264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500010902.mm.1 NM_011223.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010902.mm.1 NM_133915.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010902.mm.1 TC0500001264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500010900.mm.1 NM_011223.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010900.mm.1 NM_133915.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010900.mm.1 TC0500001264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500010913.mm.1 NM_011223.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010913.mm.1 NM_133915.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500010913.mm.1 TC0500001264.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0