This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444360_g1
See other ATF6B GE Assays ›
Gene Symbol:
Atf6b
Gene Name:
activating transcription factor 6 beta
Gene Aliases:
AA617266, ATF6beta, Creb-rp, Crebl1
Chromosome Location:
Chr.17: 34647146 - 34655074 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444360_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atf6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017406.4 NP_059102.2 1 - 2 174 64
GenBank mRNA AK090378.1 - 1 - 2 121 64
AK143765.1 - 1 - 2 107 64
AK146671.1 - 1 - 2 141 64
AK159169.1 - 1 - 2 107 64
AK170027.1 - 1 - 2 107 64
BC013534.1 - 1 - 2 174 4239106 64
BC052635.1 - 1 - 2 109 6395626 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000643.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atf6b

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
response to unfolded protein
endoplasmic reticulum unfolded protein response
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
DNA binding
transcription factor activity, sequence-specific DNA binding
cAMP response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700006271.mm.1 NM_017406.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700006271.mm.1 TC1700000643.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006274.mm.1 NM_017406.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700006274.mm.1 TC1700000643.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700006276.mm.1 NM_017406.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700006276.mm.1 TC1700000643.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 424463 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0