This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00442308_m1
See other SLC26A4 GE Assays ›
Gene Symbol:
Slc26a4
Gene Name:
solute carrier family 26, member 4
Gene Aliases:
Pds, pendrin
Chromosome Location:
Chr.12: 31519814 - 31560637 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00442308_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc26a4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011867.3 NP_035997.1 10 - 11 1481 95
GenBank mRNA AF167411.1 - 10 - 11 1482 95
BC140995.1 - 9 - 10 1329 9055607 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001595.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc26a4

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
chloride transport
regulation of pH
sulfate transport
organ morphogenesis
inorganic anion transport
iodide transport
regulation of protein localization
transmembrane transport
anion transmembrane transport
chloride transmembrane transport
secondary active sulfate transmembrane transporter activity
anion transmembrane transporter activity
chloride transmembrane transporter activity
iodide transmembrane transporter activity
sulfate transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200011070.mm.1 NM_011867.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011070.mm.1 NM_011867.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200011070.mm.1 TC1200001595.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0