This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00440839_m1
See other PRKACB GE Assays ›
Gene Symbol:
Prkacb
Gene Name:
protein kinase, cAMP dependent, catalytic, beta
Gene Aliases:
Pkacb
Chromosome Location:
Chr.3: 146729579 - 146812946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00440839_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkacb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164198.1 NP_001157670.1 4 - 5 586 81
NM_001164199.1 NP_001157671.1 4 - 5 459 81
NM_001164200.1 NP_001157672.1 4 - 5 551 81
NM_011100.4 NP_035230.1 4 - 5 590 81
GenBank mRNA AK048319.1 - 4 - 5 582 81
AK139419.1 - 4 - 5 331 81
AK148728.1 - 4 - 5 400 81
AK150122.1 - 4 - 5 552 81
AK150134.1 - 4 - 5 552 81
AK163585.1 - 4 - 5 364 81
BC054533.1 - 4 - 5 574 5704796 81
J02626.1 - 4 - 5 519 81
JN189786.1 - 4 - 5 402 81
JN189787.1 - 4 - 5 351 81
JN189788.1 - 4 - 5 326 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003106.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkacb

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
protein phosphorylation
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
phosphorylation
negative regulation of meiotic cell cycle
regulation of protein processing
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_001164198.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_001164198.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_001164199.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_001164199.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_001164200.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_001164200.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_011100.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 NM_011100.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300024450.mm.1 TC0300003106.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0