This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00439308_m1
See other HBEGF GE Assays ›
Gene Symbol:
Hbegf
Gene Name:
heparin-binding EGF-like growth factor
Gene Aliases:
AW047313, Dtr, Dts, Hegfl
Chromosome Location:
Chr.18: 36504927 - 36515805 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00439308_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hbegf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010415.2 NP_034545.1 4 - 5 844 87
GenBank mRNA AK145548.1 - 4 - 5 813 87
AK160575.1 - 4 - 5 814 87
AK164309.1 - 4 - 5 845 87
BC089607.1 - 4 - 5 813 6751441 87
L07264.1 - 4 - 5 777 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001193.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hbegf

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
blastocyst growth
epidermal growth factor receptor signaling pathway
regulation of heart contraction
cell migration
positive regulation of cell growth
positive regulation of cell migration
wound healing, spreading of epidermal cells
positive regulation of smooth muscle cell proliferation
negative regulation of elastin biosynthetic process
positive regulation of keratinocyte migration
positive regulation of protein kinase B signaling
positive regulation of wound healing
epidermal growth factor receptor binding
growth factor activity
heparin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0