This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00436905_m1
See other SLC6A6 GE Assays ›
Gene Symbol:
Slc6a6
Gene Name:
solute carrier family 6 (neurotransmitter transporter, taurine), member 6
Gene Aliases:
AA589629, C80501, Taut
Chromosome Location:
Chr.6: 91684044 - 91759063 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00436905_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc6a6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009320.4 NP_033346.2 10 - 11 1504 66
GenBank mRNA AK143510.1 - 10 - 11 1468 66
BC015245.1 - 10 - 11 1441 4242193 66
L03292.2 - 10 - 11 1476 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001092.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc6a6

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

beta-alanine transport
amino acid transmembrane transport
transport
neurotransmitter transport
taurine transport
sodium ion transmembrane transport
organic acid transmembrane transport
beta-alanine transmembrane transporter activity
neurotransmitter:sodium symporter activity
taurine transmembrane transporter activity
taurine:sodium symporter activity
symporter activity
taurine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600008559.mm.1 NM_009320.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600008559.mm.1 NM_009320.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600008559.mm.1 TC0600001092.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600008558.mm.1 NM_009320.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600008558.mm.1 NM_009320.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600008558.mm.1 TC0600001092.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0