This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00433178_g1
See other F8 GE Assays ›
Gene Symbol:
F8
Gene Name:
coagulation factor VIII
Gene Aliases:
Cf-8, Cf8, FVIII
Chromosome Location:
Chr.X: 75172715 - 75383525 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00433178_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161373.1 NP_001154845.1 16 - 17 5916 74
NM_001161374.1 NP_001154846.1 16 - 17 5916 74
NM_007977.2 NP_032003.2 17 - 18 6126 74
GenBank mRNA BC138347.1 - 16 - 17 5797 8861367 74
BC145245.1 - 16 - 17 5797 9053797 74
L05573.1 - 17 - 18 6124 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002472.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene F8

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
acute-phase response
blood coagulation
blood coagulation, intrinsic pathway
hemostasis
platelet activation
oxidation-reduction process
serine-type endopeptidase activity
copper ion binding
oxidoreductase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 NM_001161373.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 NM_001161373.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 NM_001161374.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 NM_001161374.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 NM_007977.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 NM_007977.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015797.mm.1 TC0X00002472.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0