This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf07278326_m1
See other TIGD3 GE Assays ›
Gene Symbol:
TIGD3
Gene Name:
tigger transposable element derived 3
Gene Aliases:
-
Chromosome Location:
Chr.14: 9159083 - 9161780 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf07278326_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIGD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005577292.1 XP_005577349.1 1 - 2 54 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005577292.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Probe spans exons

Panther Classification:

Molecular Function -

viral or transposable element protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header