Assay Details
Gene Symbol: PADI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544637.2 | XP_005544694.1 | 5 - 6 | 568 | 66 | |
XM_015442010.1 | XP_015297496.1 | 5 - 6 | 1556 | 66 | ||
XM_015442015.1 | XP_015297501.1 | 5 - 6 | 568 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
