Assay Details
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561540.2 | XP_005561597.1 | 5 - 6 | 1629 | 91 | |
XM_005561542.2 | XP_005561599.1 | 5 - 6 | 1628 | 91 | ||
XM_005561544.2 | XP_005561601.1 | 5 - 6 | 1627 | 91 | ||
XM_005561548.2 | XP_005561605.1 | 5 - 6 | 1627 | 91 | ||
XM_005561549.2 | XP_005561606.1 | 5 - 6 | 1627 | 91 | ||
XM_015454322.1 | XP_015309808.1 | 6 - 7 | 1530 | 91 | ||
XM_015454323.1 | XP_015309809.1 | 4 - 5 | 1133 | 91 | ||
XM_015454324.1 | XP_015309810.1 | 5 - 6 | 1562 | 91 | ||
XM_015454325.1 | XP_015309811.1 | 4 - 5 | 1446 | 91 | ||
GenBank mRNA | AB070000.1 | - | 1795 | 91 | ||
AB168202.1 | - | 6 - 7 | 1442 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
