This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04366269_m1
See other MRO GE Assays ›
Gene Symbol:
MRO
Gene Name:
maestro
Gene Aliases:
-
Chromosome Location:
Chr.18: 30894786 - 30938828 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04366269_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005586679.2 XP_005586736.1 4 - 5 478 63
XM_005586680.2 XP_005586737.1 7 - 8 1014 63
XM_005586681.2 XP_005586738.1 6 - 7 708 63
XM_005586682.2 XP_005586739.1 4 - 5 427 63
XM_005586684.2 XP_005586741.1 6 - 7 971 63
XM_005586685.2 XP_005586742.1 4 - 5 543 63
XM_005586686.2 XP_005586743.1 5 - 6 849 63
XM_015439980.1 XP_015295466.1 5 - 6 570 63
XM_015439981.1 XP_015295467.1 5 - 6 665 63
XM_015439982.1 XP_015295468.1 3 - 4 3075 63
XM_015439983.1 XP_015295469.1 4 - 5 420 63
XM_015439984.1 XP_015295470.1 2 - 3 2979 63
GenBank mRNA AB064991.1 - 5 - 6 510 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0