This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04355573_m1
See other SEPTIN11 GE Assays ›
Gene Symbol:
SEPTIN11
Gene Name:
septin 11
Gene Aliases:
SEPT11
Chromosome Location:
Chr.5: 57460995 - 57551262 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04355573_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEPTIN11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285239.1 NP_001272168.1 2 - 3 234 87
XM_015450061.1 XP_015305547.1 2 - 3 335 87
XM_015450062.1 XP_015305548.1 2 - 3 335 87
XM_015450063.1 XP_015305549.1 2 - 3 309 87
XM_015450064.1 XP_015305550.1 2 - 3 335 87
XM_015450065.1 XP_015305551.1 2 - 3 335 87
GenBank mRNA AB169689.1 - 2 - 3 234 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Panther Classification:

Molecular Function -

cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0