Assay Details
Gene Symbol: GPR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015448419.1 | XP_015303905.1 | 1 | 349 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Gene Symbol: GPR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015448419.1 | XP_015303905.1 | 1 | 349 | 133 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |