This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02845420_m1
See other XRCC4 GE Assays ›
Gene Symbol:
XRCC4
Gene Name:
X-ray repair cross complementing 4
Gene Aliases:
-
Chromosome Location:
Chr.6: 81347732 - 81640933 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02845420_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XRCC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284064.1 NP_001270993.1 5 - 6 683 140
GenBank mRNA AB169087.1 - 5 - 6 683 140
AB179303.1 - 5 - 6 683 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0